missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%

Product Code. 11384187
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384187 50 g 50g
11394187 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384187 Supplier Thermo Scientific Alfa Aesar Supplier No. 036674.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55-55-0

4-(Methylamino)phenol sulfate is a photographic developer used in black and white photography. It is also used as tone developers in developing agents. Further, it is used in oxidative hair dye formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-55-0
MDL Number MFCD00013140
Synonym Bis(4-hydroxy-n-methylanilinium) sulfate; Metol

Specifications

Melting Point ∼260°C (decomposition)
Color Cream to White
Quantity 50 g
UN Number UN2811
Beilstein 3919382
Sensitivity Air and light sensitive
Merck Index 14,6017
Solubility Information Soluble in alcohol and ether. Slightly soluble in water and ethanol. Insoluble in diethyl ether.
Formula Weight 344.38
Percent Purity 99.0 to 101.5%
Grade ACS
Physical Form Powder
Chemical Name or Material 4-(Methylamino)phenol sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-(Methylamino)phenol sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: 4-(Methylamino)phenol sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.