missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzoic Acid Ethyl Ester, TRC

Product Code. 30479373
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479373 100 mg 100mg
30481049 50 mg 50mg
30468150 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479373 Supplier Toronto Research Chemicals Supplier No. TRCM285670100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10541-82-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
Synonym 4-Ethoxycarbonyl-N-methylaniline, Ethyl 4-(Methylamino)benzoate, Ethyl p-(Methylamino)benzoate, NSC 265435
IUPAC Name ethyl 4-(methylamino)benzoate
SMILES CCOC(=O)c1ccc(NC)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H13NO2/c1-3-13-10(12)8-4-6-9(11-2)7-5-8/h4-7,11H,3H2,1-2H3
Formula Weight 179.0946
Chemical Name or Material 4-(Methylamino)benzoic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.