missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)benzoic acid, 97%

Product Code. 11409633
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409633 100 g 100g
11489623 5 g 5g
11499623 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409633 Supplier Thermo Scientific Alfa Aesar Supplier No. A12426.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10541-83-0 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-(Methylamino)benzoic acid important for the preparation of other pharmaceutical products, dyes, flavors, and preservatives. .For the preparation of medicines and other fine chemicals.

Solubility
Slightly soluble. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10541-83-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002535
InChI Key ZVIDMSBTYRSMAR-UHFFFAOYSA-N
Synonym 4-methylamino benzoic acid, benzoic acid, 4-methylamino, p-methylaminobenzoic acid, n-methyl-4-aminobenzoic acid, 4-n-methylamino benzoic acid, n-methyl-4-aminobenzoate, benzoic acid, p-methylamino, 4-methylamino-benzoic acid, p-methylamino benzoic acid, p-n-methylamino benzoic acid
PubChem CID 66345
ChEBI CHEBI:7308
IUPAC Name 4-(methylamino)benzoic acid
SMILES CNC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 158°C to 160°C
Quantity 100 g
Beilstein 2082805
Solubility Information Slightly soluble. Soluble in alcohol.
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 4- (Methylamino)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.