missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl Mebendazole, TRC

Product Code. 30471084
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471084 50 mg 50mg
30481184 250 mg 250mg
30482239 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471084 Supplier Toronto Research Chemicals Supplier No. TRCM32015550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31545-31-0
Molecular Formula C17 H15 N3 O3
Molecular Weight (g/mol) 309.32
Synonym Methyl [5-(4-Methylbenzoyl)-1H-benzimidazol-2-yl]carbamate, Mebendazole Imp. F (EP)
IUPAC Name methyl N-[5-(4-methylbenzoyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(C)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H15N3O3/c1-10-3-5-11(6-4-10)15(21)12-7-8-13-14(9-12)19-16(18-13)20-17(22)23-2/h3-9H,1-2H3,(H2,18,19,20,22)
Formula Weight 309.1113
Chemical Name or Material Methyl [5-(4-Methylbenzoyl)-1H-benzimidazol-2-yl]carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.