missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-L-phenylalanine, 98%, Thermo Scientific™

Product Code. 15463557
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15463557

Brand: Thermo Scientific Alfa Aesar H26905.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in absorption liquids on a technical scale for removing acid components such as carbon dioxide and hydrogen sulfide from gases. 4-Methyl-L-phenylalanine are used in buffers to maintain the pH of aqueous liquids at a constant level.

Solubility
Sparingly soluble (0.014 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1991-87-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00038575
InChI Key DQLHSFUMICQIMB-VIFPVBQESA-N
Synonym 4-methyl-l-phenylalanine, l-4-methylphenylalanine, 4-methylphenylalanine, h-phe 4-me-oh, 2s-2-amino-3-4-methylphenyl propanoic acid, phenylalanine, 4-methyl, s-2-amino-3-p-tolyl propanoic acid, l-4-me-phe-oh, l-4-methylphe, 4-methyl-l-phe-oh
PubChem CID 151513
IUPAC Name (2S)-2-amino-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

Solubility Information Sparingly soluble (0.014g/L) (25°C).
Quantity 250 mg
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 4-Methyl-L-phenylalanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.