missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl Histamine Dihydrochloride, TRC

Product Code. 30477007
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477007 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477007 Supplier Toronto Research Chemicals Supplier No. TRCM312015500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methyl Histamine Dihydrochloride
CAS 36376-47-3
Molecular Formula C6 H11 N3 . 2 Cl H
Quantity 500 mg
Synonym 4-Methyl-1H-imidazole-5-ethanamine, 5-Methyl-1H-imidazole-4-ethanamine Hydrochloride, 2-(5-Methyl-4-imidazolyl)ethylamine Dihydrochloride, 4-(2-Aminoethyl)-5-methylimidazole Dihydrochloride, 5-Methylhistamine Dihydrochloride, 5(4)-Methylhistamine Dihydrochloride, 5-(2-Aminoethyl)-4-
InChI Formula InChI=1S/C6H11N3.2ClH/c1-5-6(2-3-7)9-4-8-5;;/h4H,2-3,7H2,1H3,(H,8,9);2*1H
SMILES Cl.Cl.Cc1[nH]cnc1CCN
IUPAC Name 2-(5-methyl-1H-imidazol-4-yl)ethanamine;dihydrochloride
Molecular Weight (g/mol) 198.09
Formula Weight 197.0487
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.