missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methyl-d3-amino)butyric-2,2,3,3,4,4-d6 Acid HCl, CDN
Specifications
Specifications
| Chemical Name or Material | 4-(Methylamino)butanoic Acid-D8 Hydrochloride |
| CAS | 1219805-36-3 |
| Molecular Formula | C5 D9 H2 N O2 . Cl H |
| Quantity | 0.05 g |
| Synonym | 4-(Methylamino)butanoic acid-D8 hydrochloride, 4-(Methylamino)butyric-d8 acid hydrochloride, 4-(Methyl-d3-amino)butyric-2,2,3,3,4,4-d6 Acid HCl |
| InChI Formula | InChI=1 S/C5H11NO2.ClH/c1-6-4-2-3-5(7)8;/h6H,2-4H2,1H3,(H,7,8);1 H/i1D3,2D2,3D2,4D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4-hexadeuterio-4-(trideuteriomethylamino)butanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 162.6627 |
| Formula Weight | 162.1121 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?