missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-D-phenylalanine, 98%, Thermo Scientific™

Product Code. 11471780
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11471780

Brand: Thermo Scientific Alfa Aesar H27518.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49759-61-7
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00152137
InChI Key DQLHSFUMICQIMB-UHFFFAOYNA-N
Synonym 4-methyl-d-phenylalanine, h-d-phe 4-me-oh, r-2-amino-3-p-tolyl propanoic acid, d-4-methylphenylalanine, d-4-me-phe-oh, 2r-2-amino-3-4-methylphenyl propanoic acid, d-phenylalanine, 4-methyl, h-p-me-phe-oh, 4-methylphenyl-d-alanine, d-4-methylphe
PubChem CID 7015862
IUPAC Name (2R)-2-amino-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(CC(N)C(O)=O)C=C1

Specifications

Quantity 1 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 4-Methyl-D-phenylalanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.