missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4, Thermo Scientific™

Product Code. 10274154
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10274154 1 g Amber Glass Bottle 1g
10285984 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274154 Supplier Thermo Scientific Maybridge Supplier No. CC62039DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910037-15-9
Molecular Formula C14H21BN2O3
Molecular Weight (g/mol) 276.14
MDL Number MFCD09065020
InChI Key PDTRXHRSCGNYFD-UHFFFAOYSA-N
Synonym 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-boronic acid, pinacol ester, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2h,3h-pyrido 3,2-b 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h,3h-pyrido 3,2-b 1,4 oxazine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-boronic acid, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-boronic acid pinacol ester, 2-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2h-pyrido 3,2-b-1,4-oxazine,3,4-dihydro-4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-methyl-7-4,4,5,5-tetra-me-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine
PubChem CID 24229653
IUPAC Name 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydropyrido[3,2-b][1,4]oxazine
SMILES CN1CCOC2=C1N=CC(=C2)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 276.14
Percent Purity 97%
Chemical Name or Material 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.