missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10387204
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387204 1 g Amber Glass Bottle 1g
10786725 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387204 Supplier Thermo Scientific Maybridge Supplier No. CC61801DA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 57153-43-2
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.263
MDL Number MFCD01850960
InChI Key MBZWBAMMUGJINB-UHFFFAOYSA-N
Synonym 4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 4-methyl-6h,7h-thieno 3,2-c pyran-4-yl acetic acid, 4h-thieno 3,2-c pyran-4-aceticacid, 6,7-dihydro-4-methyl, 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 2-4-methyl-6,7-dihydro-4h-thiopheno 3,2-c pyran-4-yl acetic acid
PubChem CID 12225898
IUPAC Name 2-(4-methyl-6,7-dihydrothieno[3,2-c]pyran-4-yl)acetic acid
SMILES CC1(C2=C(CCO1)SC=C2)CC(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material (4-Methyl-6, 7-dihydro-4H-thieno[3, 2-c]pyran-4-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.