missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-6-(2,4,4-trimethylpentyl)-2H-pyran-2-one, TRC

Product Code. 30481070
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481070 500 mg 500mg
30466506 1 g 1g
30464625 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481070 Supplier Toronto Research Chemicals Supplier No. TRCM331260500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50650-75-4
Molecular Formula C14 H22 O2
Molecular Weight (g/mol) 222.32
Synonym USP Piroctone Related Compound B, Piroctone USP Related Compound B
IUPAC Name 4-methyl-6-(2,4,4-trimethylpentyl)pyran-2-one
SMILES CC(CC1=CC(=CC(=O)O1)C)CC(C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H22O2/c1-10-6-12(16-13(15)8-10)7-11(2)9-14(3,4)5/h6,8,11H,7,9H2,1-5H3
Formula Weight 222.162
Chemical Name or Material 4-methyl-6-(2,4,4-trimethylpentyl)-2H-pyran-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.