missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-thiazoleethanol, 98%

Product Code. 11470074
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470074 100 g 100g
11460074 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470074 Supplier Thermo Scientific Alfa Aesar Supplier No. A13567.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-00-8 | C6H9NOS | 143.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-00-8
Molecular Formula C6H9NOS
Molecular Weight (g/mol) 143.204
MDL Number MFCD00005339
InChI Key BKAWJIRCKVUVED-UHFFFAOYSA-N
Synonym 4-methyl-5-thiazoleethanol, sulfurol, 5-2-hydroxyethyl-4-methylthiazole, hemineurine, 2-4-methylthiazol-5-yl ethanol, 4-methyl-5-thiazolethanol, 5-thiazoleethanol, 4-methyl, 2-4-methyl-1,3-thiazol-5-yl ethanol, 4-methyl-5-hydroxyethylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethan-1-ol
PubChem CID 1136
ChEBI CHEBI:17957
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethanol
SMILES CC1=C(SC=N1)CCO

Specifications

Density 1.196
Boiling Point 135°C to 137°C (7 mmHg)
Flash Point 142°C (287°F)
Refractive Index 1.55
Quantity 100 g
Beilstein 114249
Merck Index 14,6126
Formula Weight 143.21
Percent Purity 98%
Chemical Name or Material 4-Methyl-5-thiazoleethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.