missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-thiazoleethanol, 98%

Product Code. 11460074
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460074 25 g 25g
11470074 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460074 Supplier Thermo Scientific Alfa Aesar Supplier No. A13567.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-00-8 | C6H9NOS | 143.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-00-8
Molecular Formula C6H9NOS
Molecular Weight (g/mol) 143.204
MDL Number MFCD00005339
InChI Key BKAWJIRCKVUVED-UHFFFAOYSA-N
Synonym 4-methyl-5-thiazoleethanol, sulfurol, 5-2-hydroxyethyl-4-methylthiazole, hemineurine, 2-4-methylthiazol-5-yl ethanol, 4-methyl-5-thiazolethanol, 5-thiazoleethanol, 4-methyl, 2-4-methyl-1,3-thiazol-5-yl ethanol, 4-methyl-5-hydroxyethylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethan-1-ol
PubChem CID 1136
ChEBI CHEBI:17957
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethanol
SMILES CC1=C(SC=N1)CCO

Specifications

Density 1.196
Boiling Point 135°C to 137°C (7 mmHg)
Flash Point 142°C (287°F)
Refractive Index 1.55
Quantity 25 g
Beilstein 114249
Merck Index 14,6126
Formula Weight 143.21
Percent Purity 98%
Chemical Name or Material 4-Methyl-5-thiazoleethanol
compliance-icons
Product Identifier
  • 4-Methyl-5-thiazoleethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methyl-5-thiazoleethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.