missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-nitroimidazole, 98%, Thermo Scientific™

Product Code. p-4229309 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11986271

Brand: Thermo Scientific Acros 440400010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14003-66-8
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD01721395
InChI Key WSYOWIMKNNMEMZ-UHFFFAOYSA-N
Synonym 4-methyl-5-nitroimidazole, 5-methyl-4-nitroimidazole, 4-methyl-5-nitro-1h-imidazole, 1h-imidazole, 4-methyl-5-nitro, imidazole, 4-methyl-5-nitro, imidazole, 5-methyl-4-nitro, unii-5pul8bw840, imidazole, 4 or 5-methyl-5 or 4-nitro, 4-methyl-5-nitro-3h-imidazole, 1h-imidazole,4-methyl-5-nitro
PubChem CID 26385
IUPAC Name 5-methyl-4-nitro-1H-imidazole
SMILES CC1=C(N=CN1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 203.2°C to 203.3°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 127.1
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Methyl-5-nitroimidazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methyl-4-nitroimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.