missing translation for 'onlineSavingsMsg'
Learn More

4'-Methyl-4-biphenylcarboxylic acid, Thermo Scientific™

Product Code. 11984991 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11984991

Brand: Thermo Scientific Acros 373220010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 720-73-0
Melting Point 252.0°C to 254.0°C
Molecular Formula C14H12O2
Linear Formula CH3(C6H4)2CO2H
Quantity 1 g
Synonym 4'-methylbiphenyl-4-carboxylic acid, 4-4-methylphenyl benzoic acid, 4'-methyl-1,1'-biphenyl-4-carboxylic acid, 4'-methyl 1,1'-biphenyl-4-carboxylic acid, 4'-methyl-4-biphenylcarboxylic acid, 4'-methyl-biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylicacid, 4'-methyl, 1,1'-biphenyl-4-carboxylic acid, 4'-methyl
InChI Key RZOCCLOTCXINRG-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
IUPAC Name 4-(4-methylphenyl)benzoic acid
Molecular Weight (g/mol) 212.25
PubChem CID 2063421
Formula Weight 212.25
Chemical Name or Material 4'-Methyl-4-biphenylcarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Methylbiphenyl-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.