missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-(trifluoromethyl)aniline, 98+%, Thermo Scientific™

Product Code. 11401007
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401007 1 g 1g
11314679 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401007 Supplier Thermo Scientific Alfa Aesar Supplier No. B23396.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65934-74-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD01631582
InChI Key JBCDCYFEJQHTTA-UHFFFAOYSA-N
Synonym 4-methyl-3-trifluoromethyl aniline, 5-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-4-methylaniline, benzenamine, 4-methyl-3-trifluoromethyl, 4-methyl-5-trifluoromethyl aniline, 4-methyl-3-trifluoromethyl-phenylamine, 4-methyl-5-trifluoromethylaniline, 3-trifluoromethyl-p-toluidine, 4-methyl-3-trifluoromethyl phenylamine
PubChem CID 2737715
IUPAC Name 4-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=C(C=C1)N)C(F)(F)F

Specifications

Density 1.24
Boiling Point 204°C
Flash Point 102°C (216°F)
Refractive Index 1.4905
Quantity 1 g
UN Number UN2810
Beilstein 6776230
Formula Weight 175.16
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.