missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrophenol, 98%

Product Code. p-7059265
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
This item is not returnable. View return policy

Product Code. 11319828

Brand: Thermo Scientific Alfa Aesar 044285.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2042-14-0 | C7H7NO3 | 153.137 g/mol

It is used as a chemical, pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2042-14-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007244
InChI Key BQEXDUKMTVYBRK-UHFFFAOYSA-N
Synonym 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol
PubChem CID 16271
IUPAC Name 4-methyl-3-nitrophenol
SMILES CC1=C(C=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 77°C to 81°C
Odor Phenol-like
Quantity 2 g
UN Number UN2446
Solubility Information Slightly soluble in water.
Formula Weight 153.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methyl-3-nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.