missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrophenol, 98%, Thermo Scientific Chemicals

Product Code. 11319828
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a chemical, pharmaceutical and OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2042-14-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007244
InChI Key BQEXDUKMTVYBRK-UHFFFAOYSA-N
Synonym 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol
PubChem CID 16271
IUPAC Name 4-methyl-3-nitrophenol
SMILES CC1=C(C=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 77°C to 81°C
Odor Phenol-like
Quantity 2 g
UN Number UN2446
Solubility Information Slightly soluble in water.
Formula Weight 153.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methyl-3-nitrophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'