missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 11995621
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 939-79-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
InChI Key KOFBNBCOGKLUOM-UHFFFAOYSA-N
Synonym 3-nitro-4-methylbenzonitrile, 3-nitro-p-tolunitrile, 4-methyl-3-nitro-benzonitrile, benzonitrile, 4-methyl-3-nitro, 4-cyano-2-nitrotoluene, 4-methyl-3-nitrobenzenecarbonitrile, 5-cyano-2-methylnitrobenzene, 2-nitro-p-toluonitrile, pubchem4817, acmc-209gtb
PubChem CID 589333
IUPAC Name 4-methyl-3-nitrobenzonitrile
SMILES CC1=C(C=C(C=C1)C#N)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 102°C to 106°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-3-nitrobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methyl-3-nitrobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'