missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-3-nitrobenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 102°C to 106°C |
Color | Brown to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Quantity | 25 g |
Formula Weight | 162.15 |
Percent Purity | 97% |
Physical Form | Crystalline Powder |
Show More |
Safety and Handling
Product Identifier
- 4-Methyl-3-nitrobenzonitrile
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'