missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzoic acid, 99%

Product Code. 11404773
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404773 500 g 500g
11494763 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404773 Supplier Thermo Scientific Alfa Aesar Supplier No. A11650.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 96-98-0 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-98-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007174
InChI Key BBEWSMNRCUXQRF-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrobenzoic acid, 3-nitro-4-methylbenzoic acid, 3-nitro-p-toluic acid, benzoic acid, 4-methyl-3-nitro, 3-nitro-para-toluic acid, m-nitro-p-toluic acid, 4-methyl-3-nitro-benzoic acid, p-toluic acid, 3-nitro, 3-nitro-4-methyl benzoic acid, benzoic acid,4-methyl-3-nitro
PubChem CID 7319
SMILES CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 188°C to 190°C
Quantity 500 g
Beilstein 1874411
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 4-Methyl-3-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.