missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitroaniline, 98%

Product Code. 11464214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464214 25 g 25g
11454214 5 g 5g
11474214 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464214 Supplier Thermo Scientific Alfa Aesar Supplier No. A12538.14
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 119-32-4 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-32-4
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007910
InChI Key GDIIPKWHAQGCJF-UHFFFAOYSA-N
Synonym 4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
PubChem CID 8390
ChEBI CHEBI:81670
IUPAC Name 4-methyl-3-nitroaniline
SMILES CC1=C(C=C(C=C1)N)[N+](=O)[O-]

Specifications

Melting Point 74°C to 79°C
Density 1.312
Flash Point 175°C (347°F)
Odor Characteristic
Quantity 25 g
UN Number UN2660
Beilstein 638984
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.