missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-heptanol, erythro + threo, 99%

Product Code. 11434257
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434257 25 g 25g
11414257 1 g 1g
11424257 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434257 Supplier Thermo Scientific Alfa Aesar Supplier No. B20357.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14979-39-6 | C8H18O | 130.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, ether, acetone. Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14979-39-6
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00004568
InChI Key BKQICAFAUMRYLZ-UHFFFAOYSA-N
Synonym 4-methyl-3-heptanol, 3-heptanol, 4-methyl, caswell no. 570a, epa pesticide chemical code 571200, 4-methyl-5-heptanol, acmc-1bqfj, dsstox_cid_21519, dsstox_rid_79763, dsstox_gsid_41519, 3-heptanol, 4-methyl-van8c
PubChem CID 26989
IUPAC Name 4-methylheptan-3-ol
SMILES CCCC(C)C(CC)O

Specifications

Density 0.83
Boiling Point 98°C to 99°C (75 mmHg)
Flash Point 59°C (138°F)
Refractive Index 1.431
Quantity 25 g
UN Number UN1987
Beilstein 1733008
Solubility Information Soluble in ethanol,ether,acetone. Slightly soluble in water.
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material 4-Methyl-3-heptanol, erythro + threo
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.