missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbaldehyde, 95%, Thermo Scientific™

Product Code. 10123364
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123364 1 g Amber Glass Bottle 1g
10526894 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123364 Supplier Thermo Scientific Maybridge Supplier No. CC62004DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-80-9
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.191
MDL Number MFCD09817506
InChI Key CVHLHSUUCRUZHB-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carbaldehyde, pubchem16127, 3,4-dihydro-7-formyl-4-methyl-2h-pyrido 3,2-b 1,4 oxazine, 4-methyl-2h,3h,4h-pyrido 3,2-b 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carbaldehyde, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazine-7-carbaldehyde, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazine-7-carboxaldehyde, 2h-pyrido 3,2-b-1,4-oxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-carboxaldehyde
PubChem CID 24229647
IUPAC Name 4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carbaldehyde
SMILES CN1CCOC2=C1N=CC(=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.