missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine, 90%, Thermo Scientific™

Product Code. 10143174
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143174 250 mg Amber Glass Bottle 250mg
10474813 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143174 Supplier Thermo Scientific Maybridge Supplier No. CC62013CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-59-4
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD09065018
InChI Key OGPGKFFHFGZOBY-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazin-7-yl methylamine, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazin-7-yl methanamine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazin-7-yl methylamine, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 1-4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazin-7-yl methanamine, 2h-pyrido 3,2-b-1,4-oxazine-7-methanamine,3,4-dihydro-4-methyl
PubChem CID 24229650
IUPAC Name (4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazin-7-yl)methanamine
SMILES CN1CCOC2=C1N=CC(=C2)CN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 179.22
Percent Purity 90%
Chemical Name or Material (4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazin-7-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.