missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10140732
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 532391-89-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key XWYHWMDBIRZPFG-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-7-carboxylic acid, 2h-1,4-benzoxazine-7-carboxylic acid, 3,4-dihydro-4-methyl, pubchem22096, 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-7-carboxyl, 4-methyl-2h,3h-benzo e 1,4-oxazaperhydroine-7-carboxylic acid
PubChem CID 2795296
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-7-carboxylic acid
SMILES CN1CCOC2=C1C=CC(=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-7-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'