missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(trifluoromethyl)benzonitrile, 98%

Product Code. 15464557
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15464557

Brand: Thermo Scientific Alfa Aesar H31904.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 261952-05-0 | C9H6F3N | 185.15 g/mol

It is used as primary and secondary reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary reagent.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261952-05-0
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD01631596
InChI Key WCZWEODOIUKVHX-UHFFFAOYSA-N
Synonym 4-methyl-2-trifluoromethyl benzonitrile, 4-methyl-2-trifluoromethyl benzenecarbonitrile, 2-cyano-5-methylbenzotrifluoride, 2-trifluoromethyl-p-tolunitrile, 4-cyano-3-trifluoromethyl toluene, benzonitrile,4-methyl-2-trifluoromethyl
PubChem CID 2775597
IUPAC Name 4-methyl-2-(trifluoromethyl)benzonitrile
SMILES CC1=CC(=C(C=C1)C#N)C(F)(F)F

Specifications

Melting Point 22°C to 28°C
Quantity 1 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 185.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.