missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(trifluoromethyl)benzoic acid, 98%

Product Code. 15445267
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15445267 Supplier Thermo Scientific Alfa Aesar Supplier No. H32534.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120985-64-0 | C9H7F3O2 | 204.15 g/mol

4-Methyl-2-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed co
TRUSTED_SUSTAINABILITY

Specifications

CAS 120985-64-0
Molecular Formula C9H7F3O2
MDL Number MFCD01631591
Quantity 1 g
Synonym 4-methyl-2-trifluoromethyl benzoic acid, 2-trifluoromethy-4-methybenzoic acid, 2-trifluoromethyl-4-methyl-benzoic acid, 4-methyl-2-trifluoromethyl-benzoic acid, 2-carboxy-5-methylbenzotrifluoride, 2-trifluoromethyl-p-toluic acid, 4-carboxy-3-trifluoromethyl toluene, 2-trifluoromethyl-4-methyl benzoic acid, 4-methyl-2-trifluoromethyl benzoicacid
Solubility Information Insoluble in water.
InChI Key PHJOGJTZXVBKJO-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C(O)=O)C(F)(F)F
IUPAC Name 4-methyl-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 204.15
PubChem CID 2775591
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-(trifluoromethyl)benzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.