missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-2-thienyl)methylamine, 97%, Thermo Scientific™

Product Code. 10177522
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177522 1 g Amber Glass Bottle 1g
10761104 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177522 Supplier Thermo Scientific Maybridge Supplier No. CC30613DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 104163-39-5
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD06657973
InChI Key CKQHNKAVFNDGMK-UHFFFAOYSA-N
Synonym 4-methylthiophen-2-yl methanamine, 4-methyl-2-thienyl methylamine, 2-thiophenemethanamine,4-methyl, 2-aminomethyl-4-methylthiophene, 4-methyl-2-aminomethyl thiophene, 4-methylthien-2-yl methylamine, 2-thiophenemethanamine, 4-methyl, 1-4-methylthiophen-2-yl methanamine, acmc-1cfbo, 4-methyl-2-thiophenemethanamine
PubChem CID 2795528
IUPAC Name (4-methylthiophen-2-yl)methanamine
SMILES CC1=CSC(=C1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 127.21
Percent Purity 97%
Chemical Name or Material (4-Methyl-2-thienyl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.