missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-thiazolecarboxaldehyde, 97%

Product Code. 10063957 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10063957 5 g Glass bottle 5g
10369197 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10063957 Supplier Thermo Scientific Acros Supplier No. 429350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13750-68-0 | C5H5NOS | 127.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13750-68-0
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.17
MDL Number MFCD00022450
InChI Key NYMCQLLAIMUVSY-UHFFFAOYSA-N
Synonym 4-methylthiazole-2-carbaldehyde, 4-methyl-2-thiazolecarboxaldehyde, 4-methyl-thiazole-2-carbaldehyde, 4-methyl-1,3-thiazole-2-carboxaldehyde, 2-thiazolecarboxaldehyde, 4-methyl, 4-methylthiazole-2-carboxaldehyde, 4-methyl-2-formylthiazole, 2-formyl-4-methyl-1,3-thiazole, 4-methyl-2-thiazole carboxaldehyde, pubchem15253
PubChem CID 4263432
IUPAC Name 4-methyl-1,3-thiazole-2-carbaldehyde
SMILES CC1=CSC(=N1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Boiling Point 90.0°C to 92.0°C (12.0 mbar)
Flash Point 67°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 127.17
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-thiazolecarboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.