missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pyrid-4-yl-1,3-thiazole-5-carbonyl chloride hydrochloride, 90%, Thermo Scientific™

Product Code. 10420574
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420574 1 g Amber Glass Bottle 1g
10758465 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10420574 Supplier Thermo Scientific Maybridge Supplier No. CC58652DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 953408-88-3
Molecular Formula C10H8Cl2N2OS
Molecular Weight (g/mol) 275.147
MDL Number MFCD09879910
InChI Key XERFFSPGBNKEDT-UHFFFAOYSA-N
Synonym 4-methyl-2-pyrid-4-yl-1,3-thiazole-5-carbonyl chloride hydrochloride, 4-methyl-2-pyridin-4-yl-1,3-thiazole-5-carbonyl chloride hydrochloride, 4-methyl-2-pyridin-4-yl-1,3-thiazole-5-carbonyl chloride-hydrogen chloride 1/1, 5-thiazolecarbonylchloride, 4-methyl-2-4-pyridinyl-, hydrochloride 1:1
PubChem CID 24229588
IUPAC Name 4-methyl-2-pyridin-4-yl-1,3-thiazole-5-carbonyl chloride;hydrochloride
SMILES CC1=C(SC(=N1)C2=CC=NC=C2)C(=O)Cl.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 275.16
Percent Purity 90%
Chemical Name or Material 4-Methyl-2-pyrid-4-yl-1, 3-thiazole-5-carbonyl chloride hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.