missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 214.22 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'