missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 98%

Product Code. 11387965
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387965 5 g 5g
11377965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387965 Supplier Alfa Aesar™ Supplier No. L08051.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22300-56-7
Molecular Formula C10H9N3O2
Molecular Weight (g/mol) 203.201
MDL Number MFCD00051653
InChI Key GQYQHAGPALBYDO-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
PubChem CID 519936
IUPAC Name 5-methyl-2-phenyltriazole-4-carboxylic acid
SMILES CC1=NN(N=C1C(=O)O)C2=CC=CC=C2

Specifications

Melting Point 200°C to 205°C
Quantity 5 g
Beilstein 177649
Formula Weight 203.2
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.