missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentyne, 97%

Product Code. 15460258
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460258 25 g 25g
15440258 1 g 1g
15450258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460258 Supplier Thermo Scientific Alfa Aesar Supplier No. H53442.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21020-27-9 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21020-27-9
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00041615
InChI Key SLMFWJQZLPEDDU-UHFFFAOYSA-N
Synonym 4-methyl-2-pentyne, 2-pentyne, 4-methyl, isopropylmethylacetylene, acmc-1co71
PubChem CID 140789
IUPAC Name 4-methylpent-2-yne
SMILES CC#CC(C)C

Specifications

Density 0.711
Boiling Point 71°C to 73°C
Flash Point −12°C (−10°F)
Linear Formula (CH3)2CHC≡CCH3
Refractive Index 1.407
Quantity 25 g
UN Number UN3295
Formula Weight 82.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-pentyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.