missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentenoic acid, 98+%

Product Code. 11465247
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465247 10 g 10g
11475247 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465247 Supplier Thermo Scientific Alfa Aesar Supplier No. B22639.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10321-71-8 | C6H9O2 | 113.14 g/mol

4-Methyl-2-pentenoic acid is used in medical research and in chemicals synthesis. It is used as a flavoring agents in foods & cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-pentenoic acid is used in medical research and in chemicals synthesis. It is used as a flavoring agents in foods & cosmetics.

Solubility
Slightly soluble in water. Soluble in acetone, ether, ethanol.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10321-71-8
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00043804
InChI Key QAOXMQCWUWZZNC-ONEGZZNKSA-M
Synonym 4-methyl-2-pentenoic acid, 4-methylpent-2-enoic acid, e-4-methylpent-2-enoic acid, 3-isopropyl acrylic acid, 2e-4-methylpent-2-enoic acid, 2-pentenoic acid, 4-methyl, unii-0qp1933ptd, 4-methylpent-2-en-1-oic acid, trans-4-methyl-2-pentenoic acid, e-4-methyl-2-pentenoic acid
PubChem CID 642039
IUPAC Name (E)-4-methylpent-2-enoic acid
SMILES CC(C)\C=C\C([O-])=O

Specifications

Density 0.959
Boiling Point 212°C
Flash Point >100°C (212°F)
Odor Unpleasant
Linear Formula (CH3)2CHCH=CHCO2H
Refractive Index 1.4485
Quantity 10 g
UN Number UN3265
Solubility Information Slightly soluble in water. Soluble in acetone,ether,ethanol.
Formula Weight 114.15
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-2-pentenoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-pentenoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methyl-2-pentenoicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.