missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentenal, 95%

Product Code. 10005513 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10005513 100 mL Glass bottle 100mL
10165510 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10005513 Supplier Thermo Scientific Acros Supplier No. 298351000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5362-56-1 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5362-56-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00800501
InChI Key RIWPMNBTULNXOH-ONEGZZNKSA-N
Synonym 4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal
PubChem CID 5916154
IUPAC Name (E)-4-methylpent-2-enal
SMILES CC(C)C=CC=O

Specifications

Acidity 15mg KOH/g max.
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8440g/mL
Boiling Point 70.0°C (75.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(CH3)CH=CHCHO
Refractive Index 1.4360 to 1.4440
Quantity 100 mL
Specific Gravity 0.844
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol,essential,oils
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentenal
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.