missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog Number 11364848
Click to view available options
Quantity:
1 L
4 x 1 L
108-10-1
C6H12O
100.161
MFCD00008938
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
7909
CHEBI:82344
4-methylpentan-2-one
CC(C)CC(=O)C
108-10-1
C6H12O
100.161
MFCD00008938
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
7909
CHEBI:82344
4-methylpentan-2-one
CC(C)CC(=O)C

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.1%, Color (APHA) 10

4-Methyl-2-pentanone is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 84°C
Color Colorless
Density 0.801 g/cm3 at 20°C
Boiling Point 117°C to 118°C
Flash Point 13°C (55°F)
Ignition Point 460°C (860°F)
Assay Percent Range >99%
Odor Mild
Refractive Index 1.396
Quantity 1 L
Beilstein 605399
Solubility Information Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water.
Formula Weight 100.16
Vapor Pressure 21 hPa (16mm Hg) at 20°C
Grade HPLC
Concentration or Composition (by Analyte or Components) 4-Methyl-2-pentanone
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone, Filtered through 0.2μ filters.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'