missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog Number 11435000
Click to view available options
Quantity:
500 mL
1 L
4 x 500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-10-1
C6H12O
100.161
MFCD00008938
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
7909
CHEBI:82344
4-methylpentan-2-one
CC(C)CC(=O)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-10-1
C6H12O
100.161
MFCD00008938
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
7909
CHEBI:82344
4-methylpentan-2-one
CC(C)CC(=O)C

4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N'-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -84°C
Color Colorless
Density 0.801 g/mL
Boiling Point 117°C to 118°C
Flash Point 13°C (55°F)
Odor Mild
Refractive Index 1.396
Quantity 500 mL
UN Number UN1245
Beilstein 605399
Merck Index 14,5207
Solubility Information Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water.
Formula Weight 100.16
Percent Purity ≥98.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'