Learn More
4-Methyl-2-pentanone, ACS, 98.5+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 108-10-1 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.161 |
MDL Number | MFCD00008938 |
InChI Key | NTIZESTWPVYFNL-UHFFFAOYSA-N |
Synonym | 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon |
PubChem CID | 7909 |
ChEBI | CHEBI:82344 |
IUPAC Name | 4-methylpentan-2-one |
SMILES | CC(C)CC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435000
|
Thermo Scientific Alfa Aesar
033346.AP |
500 mL |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397668
|
Thermo Scientific Alfa Aesar
033346.K2 |
1 L |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445000
|
Thermo Scientific Alfa Aesar
033346.AY |
4 x 500 mL |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N'-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-10-1 | |
100.161 | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
7909 | |
4-methylpentan-2-one |
C6H12O | |
MFCD00008938 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CHEBI:82344 | |
CC(C)CC(=O)C |
Specifications
108-10-1 | |
Colorless | |
117°C to 118°C | |
Mild | |
1.396 | |
UN1245 | |
14,5207 | |
Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water. | |
CC(C)CC(=O)C | |
100.161 | |
CHEBI:82344 | |
≥98.5% | |
Liquid |
-84°C | |
0.801 g/mL | |
13°C (55°F) | |
C6H12O | |
MFCD00008938 | |
605399 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
7909 | |
100.16 | |
ACS | |
4-Methyl-2-pentanone |
Safety and Handling
GHS H Statement
H225-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
H225-H319-H332-H335-H351
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ISOBUTYL KETONE
missing translation for 'einecsNumber' : 203-550-1
missing translation for 'rtecsNumber' : SA9275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only