missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS, 98.5+%

Product Code. 11397668
Change view
Click to view available options
Quantity:
500 mL
1 L
4 x 500 mL
Unit Size:
1 L
2000mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397668 1 L 1 L
11435000 500 mL 500mL
11445000 4 x 500 mL 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397668 Supplier Thermo Scientific Alfa Aesar Supplier No. 033346.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-10-1 | C6H12O | 100.161 g/mol

4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N'-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.801 g/mL
Boiling Point 117°C to 118°C
Flash Point 13°C (55°F)
Odor Mild
Refractive Index 1.396
Quantity 1 L
UN Number UN1245
Beilstein 605399
Merck Index 14,5207
Solubility Information Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water.
Formula Weight 100.16
Percent Purity ≥98.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methylisobutyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.