missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, 99+%, electronic use grade, residue free

Product Code. 10714363 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10714363

Brand: Thermo Scientific Acros 327920025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-10-1 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Acidity 0.003% max.
CAS Min % 99.0
CAS Max % 100.0
Melting Point -84.0°C
Density 0.8000g/mL
Boiling Point 117.4°C
Flash Point 14°C
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2COCH3
Refractive Index 1.3950 to 1.3970
Quantity 2.5 L
Beilstein 01, 691
Residue after Evaporation 0.001% max.
Merck Index 15, 5253
Specific Gravity 0.8
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride and acetone
Formula Weight 100.16
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.