missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanol, 99+%

Product Code. 10155303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
500mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10155303 10 L Plastic drum 10 L
10685192 5 mL Glass bottle 5mL
10725041 500 mL Glass bottle 500mL
10143290 1 L Glass bottle 1 L
10214720 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10155303 Supplier Thermo Scientific Acros Supplier No. 149380100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-11-2 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-11-2
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004550
InChI Key WVYWICLMDOOCFB-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol
PubChem CID 7910
IUPAC Name 4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -90.0°C
Color Colorless
Density 0.8000g/mL
Boiling Point 132.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic drum
Linear Formula (CH3)2CHCH2CH(OH)CH3
Refractive Index 1.4100 to 1.4120
Quantity 10 L
Beilstein 01, 410
Specific Gravity 0.8
Solubility Information Solubility in water: 20g/L. Other solubilities: misscible with most common organic solvents
Formula Weight 102.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-pentanol
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.