missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-nitrophenol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-2-nitrophenol |
| CAS | 119-33-5 |
| Molecular Formula | C7 H7 N O3 |
| Quantity | 5 g |
| Synonym | Phenol, 4-methyl-2-nitro-, p-Cresol, 2-nitro- (7CI,8CI), 4-Methyl-2-nitrophenol, 2-Nitro-4-methylphenol, 2-Nitro-p-cresol, 4-Hydroxy-3-nitrotoluene, NSC 5387, NSC 66511, o-Nitro-p-methylphenol |
| InChI Formula | InChI=1S/C7H7NO3/c1-5-2-3-7(9)6(4-5)8(10)11/h2-4,9H,1H3 |
| SMILES | Cc1ccc(O)c(c1)[N+](=O)[O-] |
| IUPAC Name | 4-methyl-2-nitrophenol |
| Molecular Weight (g/mol) | 153.14 |
| Formula Weight | 153.0426 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction