missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitrophenol, 97%, Thermo Scientific Chemicals

Product Code. 11479852
Click to view available options
Quantity:
25 g
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 32°C to 35°C
Density 1.24
Boiling Point 125°C (22 mmHg)
Flash Point 108°C (226°F)
Odor Phenol-like
Quantity 500 g
Beilstein 1868022
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-nitrophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'