missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitroaniline, 98+%

Product Code. 11470264
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470264 2500 g 2500g
11450264 100 g 100g
11460264 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470264 Supplier Thermo Scientific Alfa Aesar Supplier No. A13666.0E
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-62-3 | C7H8N2O2 | 152.15 g/mol

4-Methyl-2-nitroaniline is used as a red azo dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-nitroaniline is used as a red azo dye.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-62-3
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007907
InChI Key DLURHXYXQYMPLT-UHFFFAOYSA-N
Synonym 2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
PubChem CID 6978
ChEBI CHEBI:66920
IUPAC Name 4-methyl-2-nitroaniline
SMILES CC1=CC=C(N)C(=C1)[N+]([O-])=O

Specifications

Melting Point 115°C to 118°C
Density 1.31
Boiling Point 169°C (21 mmHg)
Flash Point 157°C (314°F)
Odor Characteristic
Quantity 2500 g
UN Number UN2660
Beilstein 879506
Solubility Information Soluble in water (0.2g/L at 20°C).
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-2-nitroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.