missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde, 97+%, Thermo Scientific™

Product Code. 10634713
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10634713 1 g Amber Glass Bottle 1g
10737115 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10634713 Supplier Thermo Scientific Maybridge Supplier No. MO00855DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 438577-61-8
Molecular Formula C12H8F3NOS
Molecular Weight (g/mol) 271.257
InChI Key TULJSSOBHAIERD-UHFFFAOYSA-N
PubChem CID 2782899
IUPAC Name 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carbaldehyde, 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbaldehyde, methyltrifluoromethylphenylthiazolecarbaldehyde, 4-methyl-2-4-trifluoromethyl-phenyl-thiazole-5-carbaldehyde, 4-methyl-2-4-trifluoromethylphenyl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxaldehyde, 5-thiazolecarboxaldehyde, 4-methyl-2-4-trifluoromethyl phenyl, 5-thiazolecarboxaldehyde,4-methyl-2-4-trifluoromethyl phenyl
Formula Weight 271.26
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.