missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(2-pyrazinyl)-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10445502
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10445502 1 g 1g
10502842 250 mg 250mg
10396394 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10445502 Supplier Thermo Scientific Maybridge Supplier No. CC19801DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 216959-92-1
Molecular Formula C9H7N3O2S
Molecular Weight (g/mol) 221.23
MDL Number MFCD00111662
InChI Key XDZXQDHRHGIPRN-UHFFFAOYSA-N
Synonym 4-methyl-2-2-pyrazinyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid, 2-5-carboxy-4-methyl-1,3-thiazol-2-yl pyrazine, 4-methyl-2-2-pyrazinyl thiazole-5-carboxylic acid, 4-methyl-2-2-pyrazinyl-thiazole-5-carboxylic acid, 4-methyl-2-pyrazin-2-yl thiazole-5-carboxylic acid, 5-carboxy-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 5-thiazolecarboxylicacid, 4-methyl-2-2-pyrazinyl, 5-thiazolecarboxylic acid, 4-methyl-2-2-pyrazinyl
PubChem CID 2776505
IUPAC Name 4-methyl-2-pyrazin-2-yl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C1=NC=CN=C1)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.24
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-(2-pyrazinyl)-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.