missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-piperazinepropionitrile, 98%, Thermo Scientific™

Product Code. 11392776
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392776 50 g 50g
11382776 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392776 Supplier Thermo Scientific Alfa Aesar Supplier No. L08905.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4491-92-3
Molecular Formula C8H15N3
Molecular Weight (g/mol) 153.229
MDL Number MFCD00051960
InChI Key XMZMOGFGIJQCHN-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl propanenitrile, 1-2-cyanoethyl-4-methylpiperazine, 3-4-methylpiperazinyl propanenitrile, acmc-1cumv, 4-methyl-1-piperazinepropionitrile, 4-methyl-1-piperazinepropanenitrile, 1-piperazinepropanenitrile, 4-methyl, n-2-cyanoethyl-n'-methylpiperazine, 3-4-methyl-piperazin-1-yl-propionitrile
PubChem CID 3050146
IUPAC Name 3-(4-methylpiperazin-1-yl)propanenitrile
SMILES CN1CCN(CC1)CCC#N

Specifications

Boiling Point 134°C to 136°C (13 mmHg)
Refractive Index 1.476
Quantity 50 g
UN Number UN3276
Beilstein 117453
Formula Weight 153.23
Percent Purity 98%
Chemical Name or Material 4-Methyl-1-piperazinepropionitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.