missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentyne, 97%

Product Code. 11367485
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367485 5 g 5g
11357485 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367485 Supplier Thermo Scientific Alfa Aesar Supplier No. L05537.06
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7154-75-8 | C6H10 | 82.15 g/mol

4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

Solubility
Insoluble in water. Soluble in common organic solvents of low polarity.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7154-75-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00039857
InChI Key OXRWICUICBZVAE-UHFFFAOYSA-N
Synonym 4-methyl-1-pentyne, 1-pentyne, 4-methyl, isobutylacetylene, isobutylethyne, 4-methylpentine, 4-methylpentyne, 4-methyl pentyne, isobutyl acetylene, 4-methyl-pentyne, 4-methyl-pent-1-yne
PubChem CID 138948
IUPAC Name 4-methylpent-1-yne
SMILES CC(C)CC#C

Specifications

Density 0.698
Boiling Point 61°C to 62°C
Flash Point −24°C (−11°F)
Refractive Index 1.393
Quantity 5 g
UN Number UN3295
Beilstein 1731077
Solubility Information Insoluble in water. Soluble in common organic solvents of low polarity.
Formula Weight 82.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-1-pentyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.