missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentene, 98+%

Product Code. 11383118
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383118 50 g 50g
11373118 10 g 10g
11393118 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11383118 Supplier Thermo Scientific Alfa Aesar Supplier No. 033029.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 691-37-2 | C6H12 | 84.16 g/mol

4-Methyl-1-pentene is used as a monomer for olefin polymerisation. The resulting polymer is poly(4-methyl-1-pentene). It can also be used to produce 1,2-diiodo-4-methyl-pentane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 691-37-2
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00008949
InChI Key WSSSPWUEQFSQQG-UHFFFAOYSA-N
Synonym 4-methyl-1-pentene, 1-pentene, 4-methyl, poly 4-methyl-1-pentene, unii-x10hrj2y7w, x10hrj2y7w, poly 4-methylpentene, isobutylethene, isohexene, 4-methylpentene-1, 1-pentene, 4-methyl-, homopolymer
PubChem CID 12724
IUPAC Name 4-methylpent-1-ene
SMILES CC(C)CC=C

Specifications

Melting Point -153°C
Color Colorless
Density 0.664 g/mL
Boiling Point 53°C to 54°C
Flash Point -32°C (-25°F)
Odor Petrol-like
Linear Formula (CH3)2CHCH2CH=CH2
Refractive Index 1.382
Quantity 50 g
UN Number UN3295
Beilstein 1731096
Solubility Information Soluble in alcohol,benzene,chloroform,petroleum ether. Insoluble in water.
Formula Weight 84.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.